Would you like to log in again?
PromoFluor, Biotin, PromoKine
Supplier: PromoKine
PK-PF700P-6-01 PF510LSS-6-01 PK-PF546-6-01 PF395LSS-6-01 PK-PF750-6-01 PF514LSS-6-01 PK-PF770-6-01 PK-PF594-6-01 PF488LSS-6-01 PK-PF680-6-01 PK-PF415-6-01 PK-PF647P-6-01 PK-PF670-6-01 PF520LSS-6-01 PK-PF568-6-01 PK-PF405-6-01 PF380LSS-6-01 PF375LSS-6-01 PK-PF532-6-01 PK-PF555P-6-01 PF500LSS-6-01 PK-PF780-6-01 PF370LSS-6-01 PK-PF488P-6-01 PK-PF633P-6-01 PF350LSS-6-01 PK-PF505-6-01
10183-128EA 419.98 USD InStock
10183-128 10183-148 10183-068 10183-046 10182-780 10183-108 10182-900 10182-802 10182-608 10182-882 10182-860 10182-586 10182-982 10182-564 10182-542 10182-520 10182-686 10182-844 10182-736 10182-758 10182-958 10182-618 10182-818 10182-652 10182-498 10183-094 10182-710
PromoFluor, Biotin, PromoKine
PromoFluor, Biotin makes it easy to detect very sensitively various proteins which are widely used in bioanalytics.
  • Water-soluble member of the B-vitamin family
  • Can be purified or detected using streptavidin
  • Ideal alternative to many commonly used fluorophores
Biotin is a water-soluble member of the B-vitamin family with a molecular weight of 244 daltons
PromoFluor Dyes show a strong light absorptionas well as high fluorescence intensity and photostability, good pH insensitivity and water solubility. The superior quality of our PromoFluor dyes and a very reasonable price making them an ideal alternative to many commonly used fluorophores.
Biotin-labeled molecules can be purified or detected using streptavidin, NeutrAvidin or monomeric avidin. The biotin-streptavidin binding - taking place under physiological conditions - is one of the the strongest noncovalent biological interactions known and biotin is a very commonly used molecule for tagging of antibodies or other proteins. Thus, this fluorescent conjugates are making it easy to detect very sensitively various proteins which are widely used in bioanalytics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty