Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
NanoJuice® Transfection Kit, MilliporeSigma
Supplier: MilliporeSigma
71900-3 71900-3
95042-078EA 378.11 USD InStock
CA95042-078 95042-078
NanoJuice® Transfection Kit, MilliporeSigma
The NanoJuice™ Transfection Kit is designed for efficient transfection of difficult-to-transfect cells, such as primary cells, Saos-2, Caco-2, HUVEC, Jurkat, Raw 264.7, HeLa, and other mammalian cells.
  • Suitable for stable and transient transfections
  • Minimal cellular toxicity
  • Compatible with both serum-containing and serum-free media
  • Allows flexible reaction conditions to optimize for specific cell types
  • Eliminates the need for media changes
  • Derived from non-animal sources
The NanoJuice™ Transfection Kit provides flexibility to fine-tune transfection conditions for efficient DNA delivery. The kit consists of two components: the NanoJuice Core Transfection Reagent and the NanoJuiceTransfection Booster.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty