Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
SP Wilmad-LabGlass Gas Collection Tubes with PTFE Plug and Side Outlet, SP Industries
Supplier: SP Industries
LG-8601T-102 LG-8601T-104
14208-440EA 673.08 USD InStock
14208-440 14205-510
SP Wilmad-LabGlass Gas Collection Tubes with PTFE Plug and Side Outlet, SP Industries
Tubes Gas Sampling Tubes
Featuring a side outlet in the body for a self sealing syringe stopper, these borosilicate glass gas collecting tubes are useful in gas chromatography or mass spectroscopy.
  • Stopcocks have PTFE plugs
For additional rubber stoppers, see ML-1940.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty