Would you like to log in again?
FUTURO™ Custom Dial Knee Strap, 3M™, National Distribution and Contracting
Supplier: National Distribution and Contracting
FUTURO™
MMM09190EN
75786-974CS 498.26 USD InStock
75786-974
FUTURO™ Custom Dial Knee Strap, 3M™, National Distribution and Contracting
Ergonomic Supports Knee Pads
Adjustable knee support with custom dial technology for personalized compression levels.
  • Turning the dial lowers the pad to apply up to 3X more pressure on the tendon
  • Adjusting the dial increases or decreases pressure without adjusting the entire strap
  • Adjustable hook and loop strap with soft edges provides for a personalized fit and added comfort
The support supplies targeted pressure on the affected knee tendon without increasing pressure around the entire leg.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty