Would you like to log in again?
FUTURO™ Sport Moisture Control Knee Support, 3M™, National Distribution and Contracting
Supplier: National Distribution and Contracting
FUTURO™
MMM45697EN MMM45699EN MMM45694EN MMM45696EN
75786-996CS 284.79 USD InStock
75786-996 75786-994 75786-992 75786-990
FUTURO™ Sport Moisture Control Knee Support, 3M™, National Distribution and Contracting
Ergonomic Supports Knee Pads
Provides support to stiff, weak or injured knees.
  • Breathable, moisture-wicking back panel for comfort
  • Dual precision straps provide a contoured, personalized fit
  • Use on right or left knee
A unique combination of neoprene-blend and breathable materials helps keep the knee warm and limber while releasing excess heat and moisture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty