Would you like to log in again?
BioPORTER® Protein Delivery Reagent, Genlantis
Supplier: Genlantis
BioPORTER®
BP509696 BP509604 BP502424 BP502401
10497-286EA 919.03 USD InStock
10497-286 10497-292 10497-290 10497-288
BioPORTER® Protein Delivery Reagent, Genlantis
Transfection Reagents
BioPORTER® protein delivery reagent is a unique lipid formulation that allows direct translocation of proteins into living cells.
  • Fast and easy protocol
  • 2X more effective than electroporation
  • Efficient delivery of a variety of proteins and peptides into a wide variety of cell types
BioPORTER Protein Delivery kits provide researchers with a quick and easy method to study protein function without the need for cloning and DNA transfection
BioPORTER kits are available as either a bulk reagent or as pre-dispensed and dried in convenient single-use QuikEase vials.
After capturing the protein, the reagent transports the lipid protein complex into the target cell. The delivered proteins retain their structure and function while leaving the transduced cells unharmed. The reagent is especially useful when studying protein function in cells that are difficult to transfect using traditional DNA transfection reagents.
Delivery information: The kit includes BioPORTER® reagent, ?-galactosidase control protein 10 µg (100 µg/mL) and FITC-antibody control protein (fluorescein-labeled goat IgG) 10 µg (100 µg/mL).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty