Would you like to log in again?
GenePORTER® Transfection Reagent Kit, Genlantis
Supplier: Genlantis
GenePORTER®
T201007H T201015H T201075H T201001 T201015 T201075 T201001S T201007 T201096
10497-770EA 2771.78 USD InStock
10497-770 10497-760 10497-756 10497-754 10497-752 10497-774 10497-772 10497-750 10497-762
GenePORTER® Transfection Reagent Kit, Genlantis
Transfection Reagents
GenePORTER® Transfection Reagent is a unique lipid formulation that allows efficient serum-free transfection of a broad spectrum of adherent and suspension cells.
  • Low cytotoxicity
  • Easy to use, robust
  • Ideal for delivery of oligonucleotides
  • High transfection efficiency in a broad spectrum of commonly transfected cell types
This reagent consists of the proprietary GenePORTER® cationic lipid together with the neutral lipid dioleoyl phosphatidylethanolamine (DOPE)
Storage: For maximum stability, store all reagents at 4 °C. If stored properly, the dried GenePORTER reagent is stable for 12 months, and the hydrated GenePORTER reagent is stable for 6 months. The Hydration Buffer and DNA Diluent are stable for 12 months.
These components provide effective DNA complexing, cytoplasmic translocation, and subsequent nuclear release of plasmid DNA and DNA oligonucleotides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty