Would you like to log in again?
GenePORTER® 2 Transfection Reagent, Genlantis
Supplier: Genlantis
GenePORTER®
T202001S T202001 T202096 T20DB05 T202075 T20DB10 T20DA05 T202007 T202015 T20DA10
10497-782EA 1239.08 USD InStock
10497-782 10497-780 10497-816 10497-814 10497-778 10497-812 10497-776 10497-810 10497-786 10497-784
GenePORTER® 2 Transfection Reagent, Genlantis
Transfection Reagents
GenePORTER® 2 Transfection Reagent is a novel lipid formulation specifically designed to provide high transfection efficiencies in a broad range of mammalian cells
  • High transfection efficiencies in difficult-to-transfect cells
  • Compatibility with serum-containing media
  • Low cytotoxicity
Storage: For maximum stability, store all reagents at 4 °C. If stored properly, the dried GenePORTER 2 reagent is stable for 12 months, and the hydrated GenePORTER 2 reagent is stable for 6 months. The Hydration Buffer and DNA Diluent are stable for 12 months.
Developed using a high-throughput transfection assay that screens a large library of cationic lipids. Its minimized interaction with serum components, make the GenePORTER 2 Reagent ideal for transfection in serum containing media.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty