Would you like to log in again?
GenePORTER® 3000 Transfection Reagent (GP3K), Genlantis
Supplier: Genlantis
GenePORTER®
T203115 T203015 T203001 T203D05 T203001S T203007
10497-792EA 541.04 USD InStock
10497-792 10497-790 10497-798 10497-788 10497-796 10497-794
GenePORTER® 3000 Transfection Reagent (GP3K), Genlantis
Transfection Reagents
GenePORTER® 3000 (GP3K) Transfection Reagent is a proprietary cationic lipid that utilizes Advanced Conjugation Enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity
  • High performance in hard-to-transfect cells
  • Fast protocol and minimal cytotoxicity
  • Stable at 4 °C for six months
This novel reagent is the result of many years of transfection expertise and applied research, and comes from a distinguished and proven pedigree of cationic lipids that provide the highest quality and performance in hard-to-transfect cells. Effective in a broad range of cell types, GP3K provides superior delivery efficiency, high transgene expression levels, and a flexible protocol, making it ideal for most delivery and protein expression experiments.
GP3K was developed for rapid same-day transfection with or without serum in a broad range of cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty