Would you like to log in again?
PrimaPure™ Human Marrow Stromal Cells (HMSC), Genlantis
Supplier: Genlantis
SH49205 SH49205K
10497-888EA 2024.71 USD InStock
10497-888 10497-886
PrimaPure™ Human Marrow Stromal Cells (HMSC), Genlantis
Cells
Human Marrow Stromal Cells (HMSC) are isolated from fetal human bone marrow and the primary HMSC provide a convenient in vitro model for investigating pathways of pluripotent cell differentiation into bone, cartilage and fat.
  • Cryopreserved at second passage
  • Each lot is functionally characterized
  • Lots from single donors to allow variability comparisons
  • Tested negative for for HIV, Hepatitis B & C, mycoplasma, bacteria, yeast, and fungi
  • Can be cultured and propagated 10 population doublings
Bone marrow stromal cells have the ability to self-renew and differentiate into cells of various connective tissue lineages, such as bone, cartilage, and fat
Each lot of PrimaPure HMSC's is tested for its ability to attach and spread on standard tissue culture grade plastic surfaces and proliferate in Marrow Stromal Cell Growth Medium.
Storage: Store cryopreserved vials in liquid nitrogen, growth medium at 4 °C in dark, subculture reagent kit at -20 °C and the reagents at 4 °C upon thawing.
The potential of HMSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue.
Delivery information: Kit contains an ampoule of cryopreserved HMSC, 500 mL of Human Marrow Stromal Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES Buffered Saline Solution, Trypsin/EDTA, and Trypsin Neutralizing Solution).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty