Would you like to log in again?
BaculoPORTER™ Transfection Reagent, Genlantis
Supplier: Genlantis
BaculoPORTER™
T701007
10497-834EA 859.37 USD InStock
10497-834
BaculoPORTER™ Transfection Reagent, Genlantis
Transfection Reagents
BaculoPORTER™ Transfection Reagent is a lipid formulation of a novel proprietary cationic lipid and dioleoyl phosphatidylethanolamine (DOPE) in a ratio targeted and optimized for the delivery of DNA into insect cells, especially Sf9 and Sf21.
  • Simple protocol and minimal cytotoxicity
  • Compatibility with diverse insect cell culture medias
  • consistently offers superior transfection efficiencies and viral titer production
BaculoPORTER Transfection Reagent coats and interacts with the DNA to form a lipoplex of lipid and DNA. The formed lipoplex is efficient at delivering the target DNA into insect cells and with minimal toxicity. This reagent allows researchers to maximize the delivery of DNA combinations including, plasmid, transfer vector and baculovirus DNA, or Bacmid DNA into insect cells to optimize expression and maximize viral titer production.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty