Would you like to log in again?
Turbo Dicer™ siRNA Generation Kit, Genlantis
Supplier: Genlantis
T520001
10497-828EA 1716.71 USD InStock
10497-828
Turbo Dicer™ siRNA Generation Kit, Genlantis
Nucleic Acid Reagents RNAi
Turbo Dicer™ siRNA Generation kit uses an ultra-active form of human recombinant dicer enzyme that can cleave more than 95% of dsRNA template into 22 bp siRNAs within two hours under optimized reaction conditions
  • Rapid two hour digestion of dsRNA in vitro
  • High yield of siRNA pool for multiple reactions
  • Effective and specific gene silencing
Turbo Dicer kit also allows to produce multiple siRNA species against target mRNA and thereby achieve effective gene silencing. Ideal for use with GeneSilencer siRNA transfection reagent. It contains sufficient reagents for generating small interfering RNAs (siRNAs) from up to five different genes and for 50 transfections in 24-well plates.
It contains sufficient reagents for preparing double stranded RNA, RNA cleavage, siRNA cleanup and transfection.
Ordering information: Recombinant Turbo Dicer Enzyme and the RNA Purification Columns can be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty