Would you like to log in again?
VWR® Freeze Spray
Supplier: VWR
61019-30
75780-352CS 217.97 USD InStock
75780-352
VWR® Freeze Spray
Freezing Sprays
Freeze Spray for electrical equipment or electronics.
  • Filtered for 100% purity
  • Leaves no residue
  • Moisture free
  • Odorless
  • Non-flammable/Non-conductive
  • Very low order of toxicity
Freeze Spray for electrical equipment or electronics.
The fastest coolant available. Five-inch polyethylene “snorkel” pinpoints freezing power exactly where needed. It cools to -60°F (-50° C) in three seconds. Saves time and eliminates water crystals caused by slower freezing methods. Pressurized liquid non-flammable gas.
A trigger-actuator valve provides controlled delivery. The pure formulation and unique valve provide superior chilling properties. Chills faster, uses less refrigerant, and maintains lower temperatures than products that use inexpensive filler material.
Certifications: DOT-1,1,1,2 Tetrafluoroethane, Non-flammable gas UN1078 DOT-E-10232, CAS No. 811-97-2.
Caution: Facilitates compliance with ENVIRONMENTAL and OSHA standards and contains no Chlorofluorocarbons or Hydrofluorocarbons. Contents under very high pressure.
Pressure at 70°F (21°C) equals 71.2 PSIG, 100°F (38°C) equals 124.3 PSIG, and 130°F (55°C) equals 198.9 PSIG. Do not puncture or incinerate container. Do not store above 120°F (49°C).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty