Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Mobile Phase Recycler, MR-100, National Scientific™
Supplier: National Scientific
BOM-LSMR100120
21506-132EA 7670.82 USD InStock
21506-132
Mobile Phase Recycler, MR-100, National Scientific™
Chromatography Systems Solvent Recyclers
The MR-100 is designed to automatically reduce solvent consumption by recycling the uncontaminated mobile phase.
  • Automatic, remote or manual modes
  • Positive and negative peak detection
  • User-defined parameters
Microprocessor-based memory stores up to ten sets of parameters, eliminating the need for reprogramming each time. The user can set a signal threshold and when no peak is detected, the recycler redirects the solvent back to the resevoir. When the signal exceeds the threshold, the recycler diverts the mobile phase into the waste reservoir, only creating waste when solvent is contaminated.
User can also set the unit to flush the system for up to 99 seconds after a peak is detected to make sure that no contaminants are sent back.
The analog signal from LC detector determine precisely when the mobile phase is recycled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty