Would you like to log in again?
innuSCRIPT Reverse Transcriptase, Analytik Jena
Supplier: Analytik Jena
845-RT-5000200 845-RT-5000050 845-RT-5000050 845-RT-5000200
10755-676EA 394.23 USD InStock
10755-676 10755-678 CA10755-676 CA10755-678
innuSCRIPT Reverse Transcriptase, Analytik Jena
Nucleic Acid Reagents RT-PCR and cDNA Reagents
innuSCRIPT Reverse Transcriptase is a genetically engineered version of MMLV reverse transcriptase. Genetic engineering has made the enzyme more specific and active in cDNA synthesis, even within a broad temperature range of 42°C to 55°C.
  • Improved specificity and activity in cDNA synthesis
  • Can be used at a wide range of temperatures (42°C - 55°C)
  • Positive test results for product sizes of 0.5 - 9 kb
  • Free of detectable endonuclease, exonuclease and RNase activity
innuSCRIPT Reverse Transcriptase is a genetically engineered version of MMLV reverse transcriptase
The enzyme tested negative for detectable endonuclease, exonuclease and RNase activity.
Genetic engineering has made the enzyme more specific and active in cDNA synthesis, even within a broad temperature range of 42°C to 55°C. Plus, innuSCRIPT Reverse Transcriptase produces higher cDNA synthesis yields than traditional enzymes, delivering excellent results for real-time PCR. innuSCRIPT Reverse Transcriptase has been tested for cDNA synthesis using various RNA starting samples and product sizes ranging from 0.5 to 9.0 kb.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty