Would you like to log in again?
High Intensity LED Stroboscopess, Nidec Shimpo
Supplier: Nidec-Shimpo
DT-365E DT-365 DT-361
75836-642EA 2333.18 USD InStock
75836-642 75836-644 75836-640
High Intensity LED Stroboscopess, Nidec Shimpo
Tachometers
The DT-360 series High-Intensity, Battery Powered LED Stroboscopes are velocity analyzing and measuring devices that are ideal for machinery process inspection.
  • Extraordinary durability with extruded aluminum, NEMA 4X construction
  • Bright LED display and simple controls for easy use
  • Long lasting, quick-charge NiMH rechargeable battery
  • LED technology greatly extends operation life and eliminates need to replace burned out bulbs
  • Capable of being sycnhronized with input signal from sensor or controller
The DT-360 Series of Stroboscopes are capable of flashing their LED lights in a synchronous frequency to the operating machinery, creating the illusion of viewing slowed or frozen images. Utilizing this phenomenon of slowing or stopping the motion with the adjusted flash rate of its LED lights, machine parts and processes may be inspected for defects, aiding in preventative maintenance programs. This LED technology extends operation due to the low energy requirement of the light diodes compared with xenon lit stroboscopes. Proper operation is assured as concern of light failure during usage, common with xenon bulbs, is eliminated.
Certifications: RoHS Approved.
Ordering information: Some accessories included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty