Would you like to log in again?
VWR® Cryovial Workstation Rack
Supplier: VWR
VWR®
T315-13VWR
75799-998CS 59.79 USD InStock
75799-998
VWR® Cryovial Workstation Rack
Racks Cryogenic Vial Racks
Constructed from sturdy polypropylene, the VWR® Cryovial Workstation Rack is compact and stackable, with strong handles for safe and easy transport.
  • Polypropylene construction
  • Holds up to 50 Cryogenic Vials of 1-5 mL
  • Autoclavable
  • Anti-skid rubber feet for benchtop stability
  • Alphanumeric indexing for easy vial identification
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty