Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tadalafil

Supplier: SELLECK CHEMICALS LLC MS
Synonyms: (6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2-methyl-2,3,12,12a-tetrahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4(6H,7H)-dione

S1512-500MG S1512-50MG S1512-100MG
101763-004EA 670 USD InStock
101763-004 101763-000 101763-002
Tadalafil
Tadalafil
Tadalafil is a PDE-5 inhibitor with IC50 of 1.8 nM.


Formula: C₂₂H₁₉N₃O₄
MW: 389.4 g/mol
MDL Number: MFCD07771966
CAS Number: 171596-29-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty