Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
VWR® Ultra-Low Temperature Chest Freezers –86 °C 230 V 50 Hz (for Export Only)
Supplier: VWR
VWRC1386V
10753-150EA 21039.62 USD InStock
10753-150
VWR® Ultra-Low Temperature Chest Freezers –86 °C 230 V 50 Hz (for Export Only)
Freezers Ultra Low Temperature Freezers
VWR® chest freezers provide versatile and reliable sample storage
  • Eye-level, easy-to-use microprocessor control panel
  • Access code to ensure setpoint security
  • Easy-to-remove air filter
  • Optional built-in chart recorder or data logger for continuous monitoring
Heavy gauge, cold-rolled steel exterior construction with a powder coat paint finish that resists chipping and rust. Convenient interior covers minimize sample exposure to outside conditions. Inner sub-lids protect bulk samples from ambient exposure during single sample retrievals. Optional CO2 or LN2 safety back-up system included for additional protection in the event of a power or mechanical failure.
Available racking solutions allow you to optimize your storage needs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty