Would you like to log in again?
Histo-Tek® Standard Cassettes, Sakura
Supplier: Sakura
9341 9345 9344 9348 9347 9340 9346 9342 9343
89199-556CS 82.5 USD InStock
89199-556 89199-566 89199-558 89199-568 89199-570 89199-572 89199-560 89199-562 89199-564
Histo-Tek® Standard Cassettes, Sakura
Autoradiography Films Cassettes
Histo-Tek® cassettes are resistant to solvent and decalcifying solutions
  • Plastic cassettes with detachable disposable lids
  • Sure-locking lid with closure guides
  • Available in 9 colors
They have unlimited applications, including identifying surgical specimens from autopsies, STAT biopsies, and frozen sections.
Packaging: Cassettes are packaged with separate compartments for lids and bases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty