Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
1 kb DNA Ladder, IBI Scientific
Supplier: IBI Scientific
IB01351
75832-840EA 74.36 USD InStock
75832-840
1 kb DNA Ladder, IBI Scientific
Electrophoresis Reagents Molecular Weight Markers
The IBI 1 kb DNA Ladder, containing 13 linear double-stranded DNA fragments is suitable for sizing double-stranded DNA from 250 to 10000 bp.
  • Ready-to-use DNA Ladder (pre-mixed with loading dye)
  • Suitable for sizing double-stranded DNA from 250 bp to 10 kb with enhanced intensities at 1 and 3 kb
  • Concentration: 2.5 µg/25 µl (Recommended loading: 0.5 µg or 5 µl)
The 1 kb DNA Ladder is a unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 13 fragments, suitable for use as molecular weight standards for electrophoresis. The enhanced intensities of the 1 and 3 kb fragments were designed for quick and easy determination of electrophoresis results.
The IBI 1 kb DNA ladder comes ready to use, no diluting necessary. Pre-mixed with 6 X loading dye made up of bromophenol blue and xylene cyanol FF.
Ships cold on blue ice and can be stored at 25 °C for 6 months, 4 °C for 12 months and −20°C for 2 years.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty