Would you like to log in again?
Vibration Meter, Extech
Supplier: Extech
VB450
75853-976EA 1486.11 USD InStock
75853-976
Vibration Meter, Extech
Vibration Meters
Convenient compact sized Vibration Meter measures vibration levels in industrial machinery.
  • Remote vibration sensor with magnetic adapter on 7.9"" (0.2m) coiled cable
  • Wide frequency range of 10Hz to 1500Hz
  • Velocity (RMS), Acceleration (Peak), and Displacement (Peak-to-Peak) measurements
  • Automatic data hold after measurement button is released
  • Auto power off and low battery indicator
The VB450 is a convenient compact sized Vibration Meter which measures vibration levels in industrial machinery. Misalignment, poor balancing, structural compromises, and other factors can cause unwanted vibration increases. VB450 features a wide 10Hz to 1500Hz frequency range and provides Displacement, Velocity, and Acceleration measurement.
Ordering information: Complete with remote sensor, magnetic mount, 9V battery, wrist strap, and soft case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty