Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Clamp Meter, Extech
Supplier: Extech
MA160 MA63 EX650 EX655 MA61
75853-954EA 497.71 USD InStock
75853-954 75853-952 75853-962 75853-956 75853-950
Clamp Meter, Extech
Electrical Meters Digital Multimeters
Our clamp meters have True RMS for accurate readings and built tough for everyday use.
  • True RMS for accurate AC measurements
  • Built-in non-contact voltage detector (NCV)
  • Low Pass Filter (LPF) for accurate measurements of variable frequency drive signals
  • Data Hold, Min/Max/Max-Min, Auto Power Off
Choice of AC Current (EX650) or fully loaded advanced AC/DC Current (EX655) model
Choice of AC Current or AC/DC Current model, both with high ±2% accuracy. Manual ranging clamp meters with three Current ranges. The 6000mA range allows for low current measurements that may be used in the automotive industry. Unique VFC (variable frequency control) mode helps troubleshoot and diagnose HVAC applications that are variable frequency driven.
Open jaw design provides quick current measurements in tight locations without breaking the circuit. True RMS provides accurate readings on noisy, distorted or non-sinusoidal waveforms. Convenient built-in Non-Contact Voltage (NCV) Detector allows quick detection of live voltages while the LED backlight, with two level brightness, and flashlight help take measurements in dimly lit environment.
Both meters feature True RMS providing accurate readings when measuring distorted or noisy waveforms and Low Impedance (LoZ) mode to eliminate false readings caused by ghost voltages. EX655 also features Low pass filter (LPF) which attenuates high frequencies to reduce noise for accurate readings and Inrush mode to capture current surges during motor startup.
Certifications: CE and UTL certified.
Ordering information: Complete with test leads, manual and batteries.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty