Would you like to log in again?
SKC High Performance Cut and Flame Resistant Sleeves, Wells Lamont
Supplier: Wells Lamont
SKC-24H SKC-18 SKC-18H SKC-24 SKC-14
10859-946PK 188.35 USD InStock
10859-946 10859-936 10859-948 10859-894 10859-950
SKC High Performance Cut and Flame Resistant Sleeves, Wells Lamont
Workwear
SKC cut and flame resistant sleeves are made from a patented combination of fibers.
  • Patented combination of fibers
  • Blousy fit
  • Elastic on both ends
  • Available in several lengths with or without a thumb hole
SKC unique knit sleeves are made from a patented combination of high performance materials and flame resistant fibers. The blousy fit of these sleeves allows for superior comfort and breathability. Elastic on both ends allows sleeve to stay in place.
Certifications: ANSI Level 3.
Ordering information: These gloves are sold by the dozen pieces.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty