Would you like to log in again?
VWR® Traceable® Decimal Stopwatch
Supplier: Avantor
90027-75
62379-229EA 153.67 USD InStock
62379-229
VWR® Traceable® Decimal Stopwatch
Timers Digital Stopwatches
Traceable® decimal stopwatches are designed for ease of operation, accuracy, and reliability.
  • Timing capacity offers 19 hours, 59 minutes, 59 seconds, and 99 hundredths
  • Quartz-crystal design is accurate to 0.0005%
  • Unit displays time of day, date, day of the week, and time-of-day alarm
  • Triple display shows cumulative splits, interval splits, and running time
Features 500 recallable memories displaying the lap, split, and running time, and also displays the fastest, slowest, and average lap time.
Tough, water-resistant ABS plastic case features tongue and groove craftsmanship and O-ring construction to seal out moisture, dust, and fumes encountered in harsh lab, plant, or field environments.
Functions as single action, time out, cumulative split, interval split, continuous roll-over timing, countdown alarm, countdown then countup alarm, countdown repeat, pacer, and stroke functions. Also used for timing enzyme tests, viscosity measurements, animal studies, and all timed lab tests.
Certifications: Compliance with ISO/IEC 17025:2005 (1750.01); calibration laboratory accredited by A2LA indicating traceability to standards provided by NIST.
Ordering information: Supplied with 40" lanyard, battery, and Traceable® certificate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty