Would you like to log in again?
Moticam® 1080 Microscope Camera, Motic Instruments
Supplier: Motic
Moticam®
1100600100831
75784-576EA 1159.2 USD InStock
75784-576
Moticam® 1080 Microscope Camera, Motic Instruments
Cameras
The Moticam® 1080 is one of Motic's most versatile microscope cameras. With the camera's on-board software, no computer is required for standard operation. Just hook up the Moticam® 1080 directly to a screen via HDMI and you're ready to capture images, make adjustments, take measurements and more. You can also capture images directly onto an SD card placed in the Moticam® 1080 unit. Should you wish to use a computer, you may do so via USB connection. Whether it’s an educational, industrial, or clinical setting, the Moticam® 1080 is ready to perform with consistent quality and convenience.
  • 8 megapixel resolution results in clear, vivid and detailed images
  • HDMI connectivity yields a super fast 60 FPS framerate
  • On-board Motic Images Plus software means no need for a computer
  • Motic Images Plus software is incuded for viewing, measurement, annotation and reporting of images
  • Comes with multiple accessories to get you started right away, including calibration slide, macro viewing tube eyepiece adapters, software, cables and more
The Moticam® 1080 brings a new level of versatility and convenience to any lab, without sacrificing performance
With the Moticam® 1080, you can still-image capture, live image record, zoom in and out, magnify, ROI, mirror, rotate, splite, freeze, cross line, and add images to a gallery. You can also make image adjustments and toggle settings. All of this can be done without the use of a computer.
Should you wish to use a computer and interact with your images in other ways, the lastest version of Motic Images Plus software is included. Just hook it up to the camera via USB or transfer the images via SD card. From there, you can measure, annotate, report and adjust them as well. No license fees or upgrade fees necessary. Just register your Moticam® online and receive free software updates as they are made available.
The Moticam® 1080 comes with everything you need to get started right away. The full package includes the camera, a cs ring adapter, HDMI cable, focusable lens, 30mm and 38mm eyepiece adapters, USB 2.0 Cable, Calibration Slide, Macro Tube, Motic Images Plus 3.0 for PC/OSX/Linux.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty