Would you like to log in again?
Cutting Mill, Variable-Speed Heavy Duty, Eberbach
E3703
75836-422EA 24775.57 USD InStock
75836-422
Cutting Mill, Variable-Speed Heavy Duty, Eberbach
Pick one of the strongest cutting mills on the market! Multiple cutting blade options allow for a wide variety of milling options. Everything you need to mill your product is included with purchase.
  • High torque
  • 7.75" diameter chamber
  • Digital controller
  • 5 year warranty
The E3703 Large Benchtop Cutting Mill features a 7.75 inch diameter sealed cutting chamber with six stationary blades and a rotating cutting head with four replaceable cutting blades that revolve at high speed to produce a shearing action which minimizes heat build-up.
Samples introduced through the hopper are swept around by the rotor until they are cut to sufficient fineness to pass through sieve. The hard, tool steel cutting edges on the blades allow milling of a wider range of materials, including plastics, animal feed, bones, cables, cardboard, electronic scrap, feed pellets, foils, food, leather, material mixtures, non-ferrous metals, paper, pharmaceutical products, refuse derived fuels, rubber, waste, wood and so much more. Optional blades are also available in 316 Stainless Steel, for use when milling potencially corrosive samples, and Tungsten Carbide, for milling harder samples.
The E3703 is designed with a machine tool type of cartridge spindle and a small powerful servo motor which allows for variable speed operation of 0 - 1740 rpm (2500 at 230V) with high torque. The chamber door comes standard with a door safety switch. The machine comes standard with mounting locations for bolting directly to a benchtop or an optional table top stand or floor stand can be purchased.
Ordering information: E3703 is supplied with 1x Plunger, 1x Hopper, 3x Sieves (0.5 mm, 1 mm and 2 mm), 1x Jar adaptor, 3x 16 oz glass jars, 1x Tool set, 1x rotating cutter head with 4x rotating blades (6 with the R6 model), 6x stationary cutting blades, and 1x brush.
Delivery information: Product ships LTL in a crate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty