Would you like to log in again?
Fetal Bovine Serum, Gold, Heat Inactivated, MP Biomedicals, LLC
Supplier: MP Biomedicals
092916849
76045-148EA 325.37 USD InStock
76045-148
Fetal Bovine Serum, Gold, Heat Inactivated, MP Biomedicals, LLC
Sera (Serum)
Fetal bovine serum has been the most widely used growth supplement for cell culture media because of its high content of embryonic growth-promoting factors
  • Supplies components that satisfy specific metabolic requirements for the culture of cells in vitro
  • Suitable for cell culture and insect cell culture
  • Source of growth promoting and survival enhancing factor
CELLect™ FBS, Gold, Heat Inactivated is used in a broad range of cell culture applications
Serum is an extremely complex mixture of many small and large biomolecules with different, physiologically balanced growth-promoting and growth-inhibiting activities. Some of the major functions of serum are to provide growth factors, hormones, attachment and spreading factors, binding proteins, lipids, and minerals.
FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty