Would you like to log in again?
Protective Cover, XS/XA Balances, Mettler Toledo
Supplier: Mettler-Toledo
11106870
11276-926EA 78 USD InStock
11276-926
Protective Cover, XS/XA Balances, Mettler Toledo
Covers
This protective cover is made out of BAREX 210 to protects balance terminal from dust, dirt and fluids.
  • Protect your terminal
  • Special material
  • Convenient operation
High resistance to a range of different chemicals. Does not interfere with visibility or operation of the terminal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty