Would you like to log in again?
SP Wilmad-LabGlass TORNADO® HPLC Reservoir, SP Industries
Supplier: SP Industries
BP-1901-001 BP-1901-005 BP-1901-010 BP-1901-002
14204-884EA 1076.06 USD InStock
14204-884 14205-816 14204-654 14206-066
SP Wilmad-LabGlass TORNADO® HPLC Reservoir, SP Industries
TORNADO® HPLC Reservoirs feature two bottom cavities; one for a TORNADO® PTFE stirrer (included), and the second for a mobile phase inlet filter.
Made of borosilicate glass, they are coated with an autoclavable red plastic that absorbs UV up to 385 nm. The mouth of reservoir has a 45 mm thread.
U.S. Patents: Des. 350,609 & 5,275,723.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty