Would you like to log in again?
SP Wilmad-LabGlass Chromatography TLC Developing Tanks, SP Industries
Supplier: SP Industries
LG-4595-112 LG-4595-120 LG-4595-102 LG-4595-110 LG-4595-100
14209-642EA 209.29 USD InStock
14209-642 14203-500 14202-924 14205-084 14203-514
SP Wilmad-LabGlass Chromatography TLC Developing Tanks, SP Industries
TLC Development Tanks
These heavy wall cylindrical borosilicate glass chromatography tanks are supplied complete with Standard Taper ground glass stoppers.
  • Available with and without ground glass stoppers
  • For use with TLC plates
These tanks are used in the development of TLC plates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty