Would you like to log in again?
HPLC Purified Activated Penetratin, MP Biomedicals
Supplier: MP Biomedicals
11PENA0500 11PENA0500
IC11PENA0500EA 797.79 USD InStock
CA76046-374 IC11PENA0500
HPLC Purified Activated Penetratin, MP Biomedicals
Nucleic Acid Reagents Transformation Reagents
Activated Penetratin™ 1 Peptide is a novel tool for peptide and siRNA delivery.
  • Successfully used to internalize covalently attached peptides, oligonucleotides, and siRNA
  • Ready for coupling to pepties or oligonucleotides containing thiol functions
  • Transfection reagent
  • Peptide
  • Lyophilized solid
The Active Penetratin™ 1 Peptide Transfection reagent features a patented 16 amino acid peptide corresponding to the third helix of the homeodomain of the Antennapedia protein (pAntp) from Drosophila melanogaster.
Penetratin™ 1 is a patented 16-amino acid peptide corresponding to the third helix of the homeodomain of antennapedeia protein. This peptide is able to translocate across biological membranes by an energy-independent mechanism (1-3) and has been used successfully to internalize covalently attached peptides (4), oligonucleotides (5) and siRNA (6). Activated Penetratin™ 1 is sold ready for coupling to peptides or oligos containing thiol (-SH) functions. Biotinylated Activated Penetratin™ 1 possesses a biotin group at the C-terminal end allowing the detection of the internalized molecule with an appropriate avidin or streptavidin reagent.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty