Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Safe Image™ Green DNA Stain, Biovision
Supplier: BioVision
Safe Image™
AB286890-1ML
76005-248EA 586.04 USD InStock
76005-248
Safe Image™ Green DNA Stain, Biovision
Stains Electrophoresis Stains
Safe Image™ Green DNA Stain
  • Safe Detection of dsDNA, ssDNA and RNA
  • Works with blue light/LED
Safe Image™ DNA Stains represents a new and safe nucleic acid stain for the visualization of double-stranded DNA (dsDNA), singlestranded DNA (ssDNA), and RNA in agarose and polyacrylamide gels. The dyes have the capability to bind to DNA and are thus developed to replace toxic Ethidium Bromide (EtBr, a potent mutagen), commonly used in gel electrophoresis for visualization of nucleic acids. Safe Image™ products are non-carcinogenic by the Ames-test. The results are negative in both the mouse marrow chromophilous erythrocyte micronucleus and mouse spermary spermatocyte chromosomal aberration tests. Safe Image™ Green DNA Stain has an Excitation Wavelength of 490 nm and Emission Wavelength of 525 nm, and its sensitivity range is between 0.2-0.6 ng. Safe Image™ Green DNA stain stains must be added to the sample before loading it to the gel. Safe Image™ Green DNA Stain will stain all nucleic acid templates (dsDNA, ssDNA and RNA) in one color. During electrophoresis, when bound to DNA, Safe Image™ Green DNA Stain emits green fluorescence.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty