Would you like to log in again?
Safe Image™ Red DNA Stain, Biovision
Supplier: BioVision
M1195-1000
76005-250EA 213.13 USD InStock
76005-250
Safe Image™ Red DNA Stain, Biovision
Safe Image™ Red DNA Stain
  • Safe detection of dsDNA, ssDNA and RNA
  • Works only under UV light
Safe Image™ DNA Stains represents a new and safe nucleic acid stain for the visualization of double-stranded DNA (dsDNA), singlestranded DNA (ssDNA), and RNA in agarose and polyacrylamide gels. The dyes have the capability to bind DNA and thus are developed to replace toxic Ethidium Bromide (EtBr, a potent mutagen), commonly used in gel electrophoresis for visualization of nucleic acids. Safe Image™ products are non-carcinogenic by the Ames-test. The results are negative in both the mouse marrow chromophilous erythrocyte micronucleus and mouse spermary spermatocyte chromosomal aberration tests. Safe Image™ Red DNA Stain has an Excitation Wavelength of 540 nm and Emission Wavelength of 630 nm, and its sensitivity range is between 0.3-0.8 ng. Safe Image™ Red DNA Stain stains must be added to the sample before loading it to the gel. Safe Image™ Red DNA Stain will stain all nucleic acid templates (dsDNA, ssDNA and RNA) in one color. Safe Image™ Red DNA Stain does have an additional excitation wavelength of around 300 nm. The other wavelength reported is for other people who may decide to use other light sources for the visualization. During electrophoresis, when bound to DNA, Safe Image™ Red DNA Stain emits red fluorescence.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty