Would you like to log in again?
Lyovapor™ L-300 Continuous Pro Modular Freeze Dryers with Pfieffer Duo 11 Vacuum Pump, Büchi
Supplier: Büchi
300PH26181124 300PH26281124 300PH26281124 300PH26181124
76046-846EA 100480.54 USD InStock
76046-846 76046-848 CA76046-846 CA76046-848
Lyovapor™ L-300 Continuous Pro Modular Freeze Dryers with Pfieffer Duo 11 Vacuum Pump, Büchi
The Lyovapor™ L-300 continuous freeze dryer offers high-quality and infinite freeze drying with complete automation.
  • Infinite-Technology™ feature allows for continuous freeze drying due to dual condensers
  • -105°C lowest condenser temperature
  • Automated cleaning
  • Sublimiation of both water and organic-based solvents
  • Advanced software for creating and managing methods, data record and reporting
The Continuous L-300 Lyovapor™ is a unique freeze dryer, providing unlimited freeze drying for the first time with BUCHI's Infinite-Technology™. Dual condensers allow for continuous sublimation of both water and organic based solvents. The Infinite-Control™ capability gives you monitoring access to all process parameters, keeping you informed by push notifications anywhere, anytime. The Pfeiffer Duo 11 vacuum pump is required and comes standard with the L-300 Continuous Lyovapor™ to allow for a fully-automated and continuous process.
Certifications: CE certified.
Ordering information: Additional configurations are available. Contact customer service for further details.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty