Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imiquimod

Supplier: MORAVEK BIOCHEMICALS MS
Synonyms: 1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine4-Amino-1-isobutyl-1H-imidazo[4,5-c]quinoline

M1822-5MG M1822-1MG
101969-726EA 738.85 USD InStock
101969-726 101969-724
Imiquimod
Imiquimod


Formula: C₁₄H₁₆N₄
MW: 240.31 g/mol
MDL Number: MFCD00866946
CAS Number: 99011-02-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty