Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

T4 RNA Ligase 2, Lucigen

Supplier: Lucigen
LR2D1132K LR2D11310K
76081-744EA 134.51 USD InStock
76081-744 76081-676
T4 RNA Ligase 2, Lucigen
Nucleic Acid Reagents Cloning Kits and Ligation
Ligate single-stranded adenylated DNA or RNA oligos to RNA targets for RNA-seq and other applications

  • Prepare cDNA libraries for small-RNA transcriptome analysis such as RNA-Seq
  • Ligate single-stranded adenylated DNA or RNA oligos to RNA targets
  • Add oligo linkers for miRNA cloning applications

T4 RNA Ligase 2, Deletion Mutant, also known as T4Rnl2(1-249), is used to ligate single-stranded adenylated DNA or RNA oligonucleotides to small RNAs for cloning or next-generation RNA sequencing

Applications Include:
RNA-Seq
miRNA cloning
Adaptor or linker ligation for RNA targets

Product Information:
Unit Definition: One unit is the amount of enzyme required to give 50% ligation of a 22-mer RNA to the preadenylated end of a 17-mer DNA when both oligos are annealed to a complementary 39-mer DNA strand in 30 minutes at 37°C under standard assay conditions.
Storage Buffer: 50% glycerol containing 50 mM Tris-HCl (pH 7.5), 0.1 M NaCl, 0.1 mM EDTA, 1 mM dithiothreitol (DTT), and 0.1% Triton® X-100.
10X Reaction Buffer: 500 mM Tris-HCl (pH 7.5), 20 mM MgCl2, and 10 mM DTT.
Activity Assay: The unit definition assay is performed in a reaction containing 50 mM Tris-HCl (pH 7.5), 2 mM MgCl2, 1 mM DTT and 0.4 µg of equimolar mix of 22-mer, 17-mer and 39-mer oligonucleotides, and varying amounts of enzyme.
Quality Control: T4 RNA Ligase 2, Deletion Mutant, is free of detectable DNA exo- and endonuclease, and RNase activities.

The preadenylated 5´ ends of DNA or RNA are ligated to the 3´ ends of RNA. Unlike the full-length enzyme, T4Rnl2(1-249) is unable to adenylate the 5´ end of the substrate in the presence of ATP. However, it can use a preactivated donor (App-DNA or App-RNA) and join it to the 3´ end of an acceptor; thus, performing the ligation reaction in the absence of ATP which prevents circularization and other undesirable bimolecular reactions.

Certifications: For research use only. Not for human or diagnostic use.

Ordering information: Contents include T4 RNA Ligase 2 Deletion Mutant enzyme and 10X Reaction Buffer.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty