Would you like to log in again?
Meridian Diagnostics Cryptococcus Antigen Latex Agglutination Test, Hardy Diagnostics
Supplier: Hardy Diagnostics
140100
76045-450EA 3809.43 USD InStock
76045-450
Meridian Diagnostics Cryptococcus Antigen Latex Agglutination Test, Hardy Diagnostics
Microbiology Test Systems Antigen-Antibody Tests Ag-AK, Latex
CALAS (Cryptococcal Antigen Latex Agglutination System) is a qualitative and semi-quantitative test system for the detection of capsular polysaccharide antigens of Cryptococcus neoformans in serum and cerebrospinal fluid (CSF).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty