Would you like to log in again?
Capacitance Substitution Box
Supplier: EISCO SCIENTIFIC LLC
PH1138
470230-408EA 58.6 USD InStock
470230-408
Capacitance Substitution Box
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for general purpose electronic work.
  • Uses 1 rotary dial with 12 steps
  • Individual values are 100pF, 220 pF, 470 pF, 2.2 nF, 4.7 nF, 10 nF, 22 nF, 47 nF, 100nF, 220 nF, 470 nF, 1 mFd (1 µF)
  • All values are + 5% accuracy and of 63V ratings
Capacitor Box, using 1 rotary dial with 12 steps. Various capacitors are selected one by one. Useful for general purpose electronic work. The unit has an easily selectable range of 12 preferred capacitors from 100 pF to 1 mFd. The individual values are 100 pF, 220 pF, 470 pF, 2.2 nF, 4.7 nF, 10 nF, 22 nF, 47 nF, 100 nF, 220 nF, 470 nF and 1 mFd (1 µF). All values of + 5% Accuracy and of 63V ratings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty