Would you like to log in again?
Neulog Thermal Conductivity Kit
Supplier: EISCO SCIENTIFIC LLC
THRKIT
470230-452KT 43.2 USD InStock
470230-452
Neulog Thermal Conductivity Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for study of thermal conductivity.
  • Study thermal conductivity of various solids
This kit allows for a qualitative and quantitative study of the thermal conductivity of different solids such as copper, brass and steel. NeuLog Temperature Sensor ( Labs Part Number 233) NOT INCLUDED.
Ordering information: NeuLog Temperature Sensor ( Labs Part Number 233) NOT INCLUDED.
Caution: For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty