Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Compositelite™ FW07, Welders Boots, Portwest
Supplier: Portwest
Compositelite™
FW07BKR47 FW07BKR39 FW07BKR43 FW07BKR38 FW07BKR42 FW07BKR41 FW07BKR48 FW07BKR45 FW07BKR46
76104-358EA 75.98 USD InStock
76104-358 76104-346 76104-356 76104-344 76104-354 76104-352 76104-340 76104-338 76104-348
Compositelite™ FW07, Welders Boots, Portwest
Shoes and Boots
Split leather boots designed for welders with a protective leather flap to guard against sparks, a quick release buckle, and dual density PU/rubber outsole.
  • Protective composite toecap
  • Puncture-resistant composite midsole
  • Water-resistant upper
  • Lightweight PU cushioned midsole
  • Slip-, fuel-, oil and heat-resistant rubber outsole
  • Ergonomic quick release buckle
  • Anti-static
Color: Black
Certifications: ASTM F2413-11
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty