Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Steelite™ Welted FW16, Safety Boots, Lace-Up, Portwest
Supplier: Portwest
Steelite™
FW16BKR48 FW16BKR40 FW16BKR45 FW16BKR44 FW16BKR43 FW16BKR41 FW16BKR46 FW16BKR39 FW16BKR42 FW16BKR47
76104-424EA 80.88 USD InStock
76104-424 76104-434 76104-422 76104-432 76104-420 76104-430 76104-418 76104-428 76104-416 76104-426
Steelite™ Welted FW16, Safety Boots, Lace-Up, Portwest
Shoes and Boots
Welted Safety Boots made from split leather upper and a rubber outsole.
  • Protective steel toecap
  • Puncture-resistant steel midsole
  • Goodyear welt construction
  • Slip-, fuel-, and oil-resistant rubber outsole
  • Heat-resistant outsole
  • Padded collar for added comfort
Padded collar and breathable lining for maximum comfort and durability. Heat- and slip-resistant outsole.
Color: Black
Certifications: ASTM F2413-11
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty