Would you like to log in again?
Heavy-Duty Inline Check Valves, Western Enterprises
Supplier: Western Enterprises
312-CVM-4F 312-CVF-4M 312-CV-4MSS 312-CVF-4MSS 312-CV-4M 312-CVM-4FSS
300007-975EA 52.92 USD InStock
300007-975 300066-734 300066-748 300066-736 300066-752 300066-750
Heavy-Duty Inline Check Valves, Western Enterprises
Welding Supplies
Heavy-duty check valves are designed for high performance in demanding applications.
  • All styles feature low/open/close pressures
  • Check valve seat material is durable Ethylene Propylene (EPDM)
  • For use in high pressure pigtail assemblies
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty