Would you like to log in again?
Flashback Arrestor Components, Western Enterprises
Supplier: Western Enterprises
312-FA-130P 312-FA-200P 312-FA-100P 312-FA-230P
300007-982EA 51.49 USD InStock
300007-982 300007-980 300007-981 300007-979
Flashback Arrestor Components, Western Enterprises
Welding Supplies
Internal check valves designed to prevent reverse flow and accidental mixing of oxygen and fuel gases.
  • Colour-coded for gas service to provide added safety
  • Torch and regulator models provide downstream and upstream protection
  • 10" maximum cutting capacity
For stopping flashbacks by preventing ignition of mixed gases and to protect equipment such as hoses, regulators, and cylinders.
Certifications: UL Listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty