Would you like to log in again?
Professional Foil/Latex Balloon Inflators, Helium, Western Enterprises
Supplier: Western Enterprises
312-RPB-62HGV 312-BR4HG 312-BR72HG
300008-010EA 170.79 USD InStock
300008-010 300066-672 300007-947
Professional Foil/Latex Balloon Inflators, Helium, Western Enterprises
Welding Supplies
  • Large hand tight connection permits easy attachment to a cylinder
  • Versatile brass push valve provides fast inflation of balloons
  • Durable, large brass tip inflates 9" to 40" latex balloons
  • Easy-to-use replaceable ribbon cutter
  • Color-coded cylinder contents gauge clearly indicates gas level
  • Tapered foil tip allows for easy insertion into foil balloons
  • Quick tie hook makes balloon tying more efficient
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty