Would you like to log in again?
REB Series Light-Duty Single Stage Regulators, Western Enterprises
Supplier: Western Enterprises
312-REB-4-3 312-REB-9-3 312-REB-1-1
300066-938EA 239.29 USD InStock
300066-938 300066-932 300066-942
REB Series Light-Duty Single Stage Regulators, Western Enterprises
Welding Supplies
For use in applications requiring a portable, lightweight, compact regulator such as in plumbing and light-duty cutting and welding.
  • Neoprene diaphragm is sensitive yet durable to maintain consistent delivery pressure
  • Dual inlet filtering system extends regulator life
  • Dual scale gauges are easy to read
  • KEL-F® seat assembly
All regulators feature a machined brass body and Neoprene diaphragm. Nominal regulator width is 3.75".
Certifications: UL Listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty