Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pig Protegrine-1 (PG-1)

Supplier: AnaSpec
AS-64819-1
103008-472EA 974.85 USD InStock
103008-472
Pig Protegrine-1 (PG-1)
Proteins and Peptides
This peptide is Protegrin-1 (PG-1) with a modified C-terminal amide. PG-1 is an 18-amino-acid beta-hairpin antimicrobial peptide found in porcine leukocytes and belongs to the cathelicidin family. PG-1 exhibits broad-spectrum activity unaffected by extracellular NaCl concentrations and is capable of inactivating numerous bacterial strains, Candida albicans, and some enveloped viruses. The efficacy is strongly dependent upon the existence of two disulfide bonds that stabilize the beta-sheet structure.
Sequence:RGGRLCYCRRRFCVCVGR-NH2 (disulfide bridge:6-15 and 8-13)
MW:2155.7 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty