Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZBTB7A IgG Polyclonal Antibody

Supplier: Boster
PB9910
76174-658EA 698.31 USD InStock
76174-658
Anti-ZBTB7A IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Zinc finger and BTB domain-containing protein 7A(ZBTB7A) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: ZBTB7A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty