Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A2]

Supplier: BioGems
05112-60-100
75841-678EA 185.21 USD InStock
75841-678
Anti-CD3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A2]
Antibodies
The 17A2 monoclonal antibody specifically reacts with the MouseT lymphocytes receptor (TCR) associated CD3 complex, resulting in cellular activation and proliferation. CD3 is expressed by thymocytes and mature lymphocytes, and contains γ, δ, and ε subunits, involved in the assembly, trafficking, and surface expression of T-cell receptor complex. The interaction between the T lymphocytes and the 17A2 antibody can be blocked by the 145-2C11 anti-CD3e antibody, demonstrating that the 17A2 antibody recognizes the CD3 ε chain.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty