Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]

Supplier: BioGems
10411-80-25
75844-324EA 127.76 USD InStock
75844-324
Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]
Antibodies
The HIT2 monoclonal antibody specifically bind to human CD38, a 45kda type II transmembrane glycoprotein. CD38 is expressed on activated T cells, plasma cells, monocytes, macrophages, dendritic cells, and thymocytes. CD38 displays cyclase and hydrolase enzymatic activity and is involved in lymphocyte adhesion, activation, and cell signaling. Its ligand is CD31.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty